CAS RN 191611-89-9
CAS RN 191611-89-9
IUPAC Name:
4-{[2-(1-methylpyrrolidin-2-yl)ethyl]sulfanyl}phenol hydrochloride
Synonyms:
4-{[2-(1-methylpyrrolidin-2-yl)ethyl]sulfanyl}phenol hydrochloride
Molecular Weight (g/mol):
273.82
Molecular Formula:
C13H20ClNOS
InChi Key:
RJSVKPQKXSMZMT-UHFFFAOYNA-N
SMILES:
Cl.CN1CCCC1CCSC1=CC=C(O)C=C1
1
–
3
of
3
results
1
SIB 1553A Hyrdrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
SIB 1553A Hyrdrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
SIB 1553A Hyrdrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.