Learn More
SIB 1553A Hyrdrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | SIB 1553A Hyrdrochloride |
| CAS | 191611-89-9 |
| Molecular Formula | C13H19NOS • HCl |
| Quantity | 100 mg |
| Synonym | 4-((2-(1-Methylpyrrolidin-2-yl)ethyl)thio)phenol Hydrochloride |
| InChI Formula | InChI=1S/C13H19NOS.ClH/c1-14-9-2-3-11(14)8-10-16-13-6-4-12(15)5-7-13;/h4-7,11,15H,2-3,8-10H2,1H3;1H |
| SMILES | OC1=CC=C(SCCC2N(C)CCC2)C=C1.Cl |
| IUPAC Name | 4-[2-(1-methylpyrrolidin-2-yl)ethylsulfanyl]phenol;hydrochloride |
| Molecular Weight (g/mol) | 237.363646 |
| Formula Weight | 273.0954 |