Learn More
SIB 1553A Hyrdrochloride, TRC
Product Code. 30410368
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410368 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30410368 Supplier Toronto Research Chemicals Supplier No. TRCS41100050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material SIB 1553A Hyrdrochloride
CAS 191611-89-9
Molecular Formula C13H19NOS • HCl
Quantity 50 mg
Synonym 4-((2-(1-Methylpyrrolidin-2-yl)ethyl)thio)phenol Hydrochloride
InChI Formula InChI=1S/C13H19NOS.ClH/c1-14-9-2-3-11(14)8-10-16-13-6-4-12(15)5-7-13;/h4-7,11,15H,2-3,8-10H2,1H3;1H
SMILES OC1=CC=C(SCCC2N(C)CCC2)C=C1.Cl
IUPAC Name 4-[2-(1-methylpyrrolidin-2-yl)ethylsulfanyl]phenol;hydrochloride
Molecular Weight (g/mol) 237.363646
Formula Weight 273.0954
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.