accessibility menu, dialog, popup

UserName

CAS RN 1904-98-9

NH2NNH2HNNNOHSOHOO

CAS RN 1904-98-9

IUPAC Name: 7H-purine-2,6-diamine; sulfuric acid
Synonyms: 2,6-diaminopurine
Molecular Weight (g/mol): 248.22
Molecular Formula: C5H8N6O4S
InChi Key: KYYLASPNTMEZTB-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC(N)=C2NC=NC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results