accessibility menu, dialog, popup

UserName

HOOFClFFF

CAS RN 186517-41-9

IUPAC Name: 3-chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
Synonyms: 3-chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 242.55
Molecular Formula: C8H3ClF4O2
InChi Key: PCNHWBAPTNAREE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(Cl)=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results