accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 17628-72-7

IUPAC Name: 2-methoxy-2-phenylethan-1-ol
Synonyms: 2-methoxy-2-phenylethanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: JDTUPLBMGDDPJS-UHFFFAOYNA-N
SMILES: COC(CO)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results