missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methoxy-2-phenylethanol, 98+%

Product Code. 11398925
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398925 250 mg 250mg
11388925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398925 Supplier Thermo Scientific Alfa Aesar Supplier No. L14044.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17628-72-7 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17628-72-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00011550
InChI Key JDTUPLBMGDDPJS-VIFPVBQESA-N
Synonym r-2-methoxy-2-phenylethanol, r---2-methoxy-2-phenylethanol, 2r-2-methoxy-2-phenylethanol, 2r-2-methoxy-2-phenyl-ethanol, r-2-phenyl-2-methoxyethanol, --, a-methoxyphenethyl alcohol, benzeneethanol, beta-methoxy-, betar
PubChem CID 6950345
IUPAC Name (2R)-2-methoxy-2-phenylethanol
SMILES COC(CO)C1=CC=CC=C1

Specifications

Density 1.05
Boiling Point 65°C (0.1 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.52
Quantity 250 mg
Beilstein 2437247
Optical Rotation −133° (c=1 in Acetone)
Formula Weight 152.19
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-Methoxy-2-phenylethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.