CAS RN 171103-04-1
CAS RN 171103-04-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Desmethylthiamethoxam, TRC
CAS: 171103-04-1 Molecular Formula: C7 H8 Cl N5 O3 S Molecular Weight (g/mol): 277.6881 Synonym: 2H-1,3,5-Oxadiazin-4-amine, 3-[(2-chloro-5-thiazolyl)methyl]-3,6-dihydro-N-nitro-,3-[(2-Chloro-5-thiazolyl)methyl]-3,6-dihydro-N-nitro-2H-1,3,5-oxadiazin-4-amine,CGA 330050,Desmethylthiamethoxam,N-Desmethylthiamethoxam IUPAC Name: N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-2,6-dihydro-1,3,5-oxadiazin-4-yl]nitramide SMILES: [O-][N+](=O)NC1=NCOCN1Cc2cnc(Cl)s2
2
Thiamethoxam-desmethyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality