CAS RN 170570-01-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-[5-(3-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Desmethyl-3-methyl Celecoxib, TRC
CAS: 170570-01-1 Molecular Formula: C17 H14 F3 N3 O2 S Molecular Weight (g/mol): 381.37 Synonym: 4-[5-(3-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide,Celecoxib Imp. A (Pharmeuropa) IUPAC Name: 4-[5-(3-methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide SMILES: Cc1cccc(c1)c2cc(nn2c3ccc(cc3)S(=O)(=O)N)C(F)(F)F