accessibility menu, dialog, popup

UserName

CAS RN 1689-82-3

ONNH

CAS RN 1689-82-3

IUPAC Name: 4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Synonyms: 4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: JTSBGMZPPPULTA-UHFFFAOYSA-N
SMILES: O=C1C=CC(C=C1)=NNC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results