CAS RN 1619994-69-2
CAS RN 1619994-69-2
IUPAC Name:
ethyl N-[6-(3-methanesulfonamido-4-methylphenyl)-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-yl]carbamate
Synonyms:
ethyl N-[6-(3-methanesulfonamido-4-methylphenyl)-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-yl]carbamate
Molecular Weight (g/mol):
404.45
Molecular Formula:
C17H20N6O4S
InChi Key:
UYBRROMMFMPJAN-UHFFFAOYSA-N
SMILES:
CCOC(=O)NC1=CC(=NN2C(C)=NN=C12)C1=CC(NS(C)(=O)=O)=C(C)C=C1
1
–
4
of
4
results
1
Bromosporine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bromosporine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Bromosporine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.