missing translation for 'onlineSavingsMsg'
Learn More

Bromosporine, TRC

Product Code. 30347759
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30347759

Brand: Toronto Research Chemicals TRCB6875005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromosporine
CAS 1619994-69-2
Molecular Formula C17H20N6O4S
Quantity 5 mg
Synonym Ethyl (3-Methyl-6-(4-methyl-3-(methylsulfonamido)phenyl)-[1,2,4]triazolo[4,3-b]pyridazin-8-yl)carbamate, Ethyl N-[3-methyl-6-[4-methyl-3-[(methylsulfonyl)amino]phenyl]-1,2,4-triazolo[4,3-b]pyridazin-8-yl]carbamate (ACI), Carbamic acid, N-(3-methyl-6-(4-methyl-3-((methylsulfonyl)amino)phenyl)-1,2,4-triazolo(4,3-b)pyridazin-8-yl)-, ethyl ester, Ethyl (3-methyl-6-(4-methyl-3-((methylsulfonyl)amino)phenyl)(1,2,4)triazolo(4,3-b)pyridazin-8-yl)carbamate
InChI Formula InChI=1S/C17H20N6O4S/c1-5-27-17(24)18-15-9-14(21-23-11(3)19-20-16(15)23)12-7-6-10(2)13(8-12)22-28(4,25)26/h6-9,22H,5H2,1-4H3,(H,18,24)
SMILES O=C(OCC)NC1=CC(=NN2C1=NN=C2C)C=3C=CC(=C(C3)NS(=O)(=O)C)C
IUPAC Name ethyl (3-methyl-6-(4-methyl-3-(methylsulfonamido)phenyl)-[1,2,4]triazolo[4,3-b]pyridazin-8-yl)carbamate
Molecular Weight (g/mol) 404.44
Formula Weight 404.13
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.