accessibility menu, dialog, popup

UserName

CH3CH3NN

CAS RN 1558-17-4

IUPAC Name: 4,6-dimethylpyrimidine
Synonyms: 4,6-dimethylpyrimidine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: LSBIUXKNVUBKRI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results