missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethylpyrimidine, 94%

Product Code. 11434053
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434053 5 g 5g
11424053 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434053 Supplier Thermo Scientific Alfa Aesar Supplier No. A10302.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1558-17-4 | C6H8N2 | 108.14 g/mol

4,6-Dimethylpyrimidine is used to produce (6-methyl-pyrimidin-4-yl)-pyruvic acid ethyl ester. It is used as pharmaceutical intermediate. It is also used in agrochemical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dimethylpyrimidine is used to produce (6-methyl-pyrimidin-4-yl)-pyruvic acid ethyl ester. It is used as pharmaceutical intermediate. It is also used in agrochemical and dyestuff field.

Solubility
Soluble in water at 20°C.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1558-17-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006116
InChI Key LSBIUXKNVUBKRI-UHFFFAOYSA-N
Synonym 4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
PubChem CID 15257
IUPAC Name 4,6-dimethylpyrimidine
SMILES CC1=CC(C)=NC=N1

Specifications

Density 0.98
Boiling Point 153°C to 155°C
Flash Point 46°C (114°F)
Odor Recognizable
Refractive Index 1.49
Quantity 5 g
UN Number UN1993
Beilstein 107068
Solubility Information Soluble in water at 20°C.
Formula Weight 108.14
Percent Purity 94%
Chemical Name or Material 4,6-Dimethylpyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.