CAS RN 15045-43-9
CAS RN 15045-43-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2,2,5,5-Tetramethyloxolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2,5,5-Tetramethyloxolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2,5,5-Tetramethyloxolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.