Learn More
2,2,5,5-Tetramethyloxolane, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2,5,5-Tetramethyloxolane |
| CAS | 15045-43-9 |
| Molecular Formula | C8 H16 O |
| Quantity | 50 mg |
| Synonym | 2,2,5,5-Tetramethyltetrahydrofuran |
| InChI Formula | InChI=1S/C8H16O/c1-7(2)5-6-8(3,4)9-7/h5-6H2,1-4H3 |
| SMILES | CC1(C)CCC(C)(C)O1 |
| IUPAC Name | 2,2,5,5-tetramethyloxolane |
| Molecular Weight (g/mol) | 128.212 |
| Formula Weight | 128.12 |