CAS RN 149647-78-9
CAS RN 149647-78-9
IUPAC Name:
N-hydroxy-N'-phenyloctanediamide
Synonyms:
vorinostat vorinostat (product) M344 compound nhnpoda vorinostat (substance) SHH zolinza suberanilohydroxamic acid SAHA saha cpd
Molecular Weight (g/mol):
264.33
Molecular Formula:
C14H20N2O3
InChi Key:
WAEXFXRVDQXREF-UHFFFAOYSA-N
SMILES:
ONC(=O)CCCCCCC(=O)NC1=CC=CC=C1
1
–
3
of
3
results
1
Tocris Bioscience™ SAHA
CAS: 149647-78-9 Molecular Formula: C14H20N2O3
2
Vorinostat, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3