CAS RN 144403-08-7
CAS RN 144403-08-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(2S)-6-Oxo-1,2-piperidinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S)-6-Oxo-1,2-piperidinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2S)-6-Oxo-1,2-piperidinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.