missing translation for 'onlineSavingsMsg'
Learn More

(2S)-6-Oxo-1,2-piperidinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester, TRC

Product Code. 30410324
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410324 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410324 Supplier Toronto Research Chemicals Supplier No. TRCO85880525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-6-Oxo-1,2-piperidinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester
CAS 144403-08-7
Molecular Formula C15H25NO5
Quantity 25 mg
InChI Formula InChI=1S/C15H25NO5/c1-14(2,3)20-12(18)10-8-7-9-11(17)16(10)13(19)21-15(4,5)6/h10H,7-9H2,1-6H3/t10-/m0/s1
SMILES CC(C)(C)OC([C@H]1N(C(CCC1)=O)C(OC(C)(C)C)=O)=O
IUPAC Name di-tert-butyl (S)-6-oxopiperidine-1,2-dicarboxylate
Molecular Weight (g/mol) 299.36
Formula Weight 299.17
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.