accessibility menu, dialog, popup

UserName

CAS RN 144-19-4

CH3CH3HOCH3H3CHO

CAS RN 144-19-4

IUPAC Name: 2,2,4-trimethylpentane-1,3-diol
Synonyms: trimethyl-1,3-pentanediol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: JCTXKRPTIMZBJT-UHFFFAOYNA-N
SMILES: CC(C)C(O)C(C)(C)CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)