Learn More
2,2,4-Trimethyl-1,3-pentanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2,4-Trimethyl-1,3-pentanediol |
| CAS | 144-19-4 |
| Molecular Formula | C8 H18 O2 |
| Quantity | 250 g |
| Synonym | 2,2,4-Trimethyl-1,3-pentanediol, 2,2,4-Trimethylpentan-1,3-diol, Nexcoat 795, NSC 6368, TMPD (alcohol), 2,2,4-Trimethyl-1,3-pentanediol, NSC 6368, Nexcoat 795, TMPD (Alcohol) |
| InChI Formula | InChI=1S/C8H18O2/c1-6(2)7(10)8(3,4)5-9/h6-7,9-10H,5H2,1-4H3 |
| SMILES | CC(C)C(O)C(C)(C)CO |
| IUPAC Name | 2,2,4-trimethylpentane-1,3-diol |
| Molecular Weight (g/mol) | 146.227 |
| Formula Weight | 146.1307 |