accessibility menu, dialog, popup

UserName

CAS RN 1429-50-1

NH3OHPHOONNPOHOHOPHOOHOPOHHOO

CAS RN 1429-50-1

IUPAC Name: [({2-[bis(phosphonomethyl)amino]ethyl}(phosphonomethyl)amino)methyl]phosphonic acid amine
Synonyms: edtmp
Molecular Weight (g/mol): 453.15
Molecular Formula: C6H23N3O12P4
InChi Key: STNSYZSNIYTNMI-UHFFFAOYSA-N
SMILES: N.OP(O)(=O)CN(CCN(CP(O)(O)=O)CP(O)(O)=O)CP(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results