missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetra(methylenephosphonic acid), TRC

Product Code. 30388384
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388384 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388384 Supplier Toronto Research Chemicals Supplier No. TRCE9183235G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylenediaminetetra(methylenephosphonic acid)
CAS 1429-50-1
Molecular Formula C6H20N2O12P4
Quantity 5 g
Synonym P,P',P'',P'''-[1,2-Ethanediylbis[nitrilobis(methylene)]]tetrakis[phosphonic Acid], EDPA, EDTMP, EDTPA, EDTPH, Chelating Agent 540, Ethylenedi(nitrilodimethylene)tetraphosphonic Acid, Ethylenediamine-N,N,N',N'-tetra(methylphosphonic Acid), Ethylenediaminetetra(methylenephosphonic Acid), Ethylenediaminetetramethylenephosphonate, N,N,N',N'-Tetrakis(phosphonomethyl)ethylenediamine, [Ethylenebis(nitrilodimethylene)]tetraphosphonic Acid
InChI Formula InChI=1S/C6H20N2O12P4/c9-21(10,11)3-7(4-22(12,13)14)1-2-8(5-23(15,16)17)6-24(18,19)20/h1-6H2,(H2,9,10,11)(H2,12,13,14)(H2,15,16,17)(H2,18,19,20)
SMILES OP(=O)(O)CN(CCN(CP(=O)(O)O)CP(=O)(O)O)CP(=O)(O)O
IUPAC Name [2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid
Molecular Weight (g/mol) 436.12
Formula Weight 435.9967
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.