accessibility menu, dialog, popup

UserName

CH3CH3SOONHOOH

CAS RN 13240-18-1

IUPAC Name: 2-(2,5-dimethylbenzenesulfonamido)acetic acid
Synonyms: (2,5-dimethylbenzenesulfonamido)acetic acid
Molecular Weight (g/mol): 243.28
Molecular Formula: C10H13NO4S
InChi Key: LQBDYUYIAIGJGX-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)C=C1)S(=O)(=O)NCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)