missing translation for 'onlineSavingsMsg'
Learn More

N-(2,5-Dimethylphenylsulfonyl)glycine, 96%, Thermo Scientific™

Product Code. 15445827
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445827 250 mg 250mg
15425827 1 g 1g
15435827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15445827 Supplier Thermo Scientific Alfa Aesar Supplier No. H33039.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13240-18-1
Molecular Formula C10H13NO4S
Molecular Weight (g/mol) 243.277
MDL Number MFCD03658020
InChI Key LQBDYUYIAIGJGX-UHFFFAOYSA-N
Synonym 2,5-dimethyl-benzenesulfonylamino-acetic acid, n-2,5-dimethylphenyl sulfonyl glycine, 2-2,5-dimethylbenzenesulfonamido acetic acid, 2,5-dimethylbenzenesulfonamidoacetic acid, n-2,5-dimethylbenzene-1-sulfonyl glycine, glycine,n-2,5-xylylsulfonyl-7ci,8ci, 2,5-dimethylbenzenesulfonamido acetic acid, 2-2,5-dimethylphenyl sulfonyl amino acetic acid, n-2,5-dimethylphenylsulfonyl glycine, 2,5-dimethylphenylsulfonylamino acetic acid
PubChem CID 757395
IUPAC Name 2-[(2,5-dimethylphenyl)sulfonylamino]acetic acid
SMILES CC1=CC(=C(C=C1)C)S(=O)(=O)NCC(=O)O

Specifications

Melting Point 130°C to 131°C
Quantity 250 mg
Formula Weight 243.28
Percent Purity 96%
Chemical Name or Material N-(2,5-Dimethylphenylsulfonyl)glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.