CAS RN 126325-47-1
IUPAC Name:
6-bromo-2-methylpyridin-3-amine
Synonyms:
6-bromo-2-methylpyridin-3-amine
Molecular Weight (g/mol):
187.04
Molecular Formula:
C6H7BrN2
InChi Key:
UBTQJSZELCWGCN-UHFFFAOYSA-N
SMILES:
CC1=C(N)C=CC(Br)=N1
1
–
1
of
1
results
1
5-Amino-2-bromo-6-methylpyridine, 98%, Thermo Scientific™
CAS: 126325-47-1 Molecular Formula: C6H7BrN2 Molecular Weight (g/mol): 187.04 MDL Number: MFCD03095086 InChI Key: UBTQJSZELCWGCN-UHFFFAOYSA-N Synonym: 5-amino-2-bromo-6-picoline,3-amino-6-bromo-2-methylpyridine,2-bromo-5-amino-6-methylpyridine,3-amino-6-bromo-2-picoline,5-amino-2-bromo-6-methylpyridine,2-bromo-5-amino-6-picoline,6-bromo-2-methyl-3-pyridylamine,6-bromo-2-methyl-pyridin-3-ylamine,2-methyl-3-amino-6-bromopyridine,pubchem1117 PubChem CID: 14418045 IUPAC Name: 6-bromo-2-methylpyridin-3-amine SMILES: CC1=C(N)C=CC(Br)=N1