Learn More
5-Amino-2-bromo-6-methylpyridine, 98%, Thermo Scientific™
Product Code. 15422089
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422089 25 g 25g
15402089 1 g 1g
15412089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15422089 Supplier Thermo Scientific Alfa Aesar Supplier No. H64756.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 126325-47-1
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD03095086
InChI Key UBTQJSZELCWGCN-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
PubChem CID 14418045
IUPAC Name 6-bromo-2-methylpyridin-3-amine
SMILES CC1=C(N)C=CC(Br)=N1
Specifications
Melting Point 110°C to 111°C
Color White
Quantity 25 g
Beilstein 6190039
Formula Weight 187.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Amino-2-bromo-6-methylpyridine