accessibility menu, dialog, popup

UserName

CAS RN 125317-39-7

CH3CH2NNH(S)OOCH3OCH3NCH3(R)(S)OH(R)OH3CO(R)H3CN(R)(R)OOH3COH(R)(R)OHHOOOHOOH(R)(R)OHHOOOHO

CAS RN 125317-39-7

IUPAC Name: bis((2R,3R)-2,3-dihydroxybutanedioic acid); methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(12S)-16-ethyl-12-(methoxycarbonyl)-1,10-diazatetracyclo[12.3.1.0³,¹¹.0⁴,⁹]octadeca-3(11),4,6,8,15-pentaen-12-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate
Synonyms: bis(L(+)-tartaric acid); vinorelbine
Molecular Weight (g/mol): 1079.12
Molecular Formula: C53H66N4O20
InChi Key: CILBMBUYJCWATM-NPJYPKOYSA-N
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CCC1=CC2CN(C1)CC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results