accessibility menu, dialog, popup

UserName

CAS RN 1240257-48-0

CH3SFFFFFBr

CAS RN 1240257-48-0

IUPAC Name: 1-(bromomethyl)-3-methyl-5-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-(bromomethyl)-3-methyl-5-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 311.11
Molecular Formula: C8H8BrF5S
InChi Key: ZRHPUHTTXXCZFI-UHFFFAOYSA-N
SMILES: CC1=CC(CBr)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results