accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 124-06-1

IUPAC Name: ethyl tetradecanoate
Synonyms: ethylmyristate
Molecular Weight (g/mol): 256.43
Molecular Formula: C16H32O2
InChi Key: MMKRHZKQPFCLLS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OCC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)