accessibility menu, dialog, popup

UserName

CAS RN 118-75-2

ClClOClOCl

CAS RN 118-75-2

IUPAC Name: tetrachlorocyclohexa-2,5-diene-1,4-dione
Synonyms: chloranil α-chloranil tetrachloroparabenzoquinone
Molecular Weight (g/mol): 245.86
Molecular Formula: C6Cl4O2
InChi Key: UGNWTBMOAKPKBL-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)