missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloro-1,4-benzoquinone, TRC

Product Code. 30425262
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30425262

Brand: Toronto Research Chemicals TRCT291355100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloranil
CAS 118-75-2
Molecular Formula C6 Cl4 O2
Quantity 100 g
Synonym p-Benzoquinone','2,3,5,6-tetrachloro- (8CI), 2,3,5,6-Tetrachloro-2,5-cyclohexadiene-1,4-dione, 1,2,4,5-Tetrachlorobenzoquinone, 2,3,5,6-Tetrachloro-1,4-(p-)-benzoquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 2,3,5,6-Tetrachloro-p-benzoquinone, 2,3,5,6-Tetrachlorobenzoquinone, 2,3,5,6-Tetrachlorocyclohexadiene-1,4-dione, 2,3,5,6-Tetrachloroquinone, Actor CL, Chloranil, Coversan, Dow Seed Disinfectant No. 5, ENT 3797, NSC 8432, Psorisan, Quinone tetrachloride, Reranil, Spergon, Spergon I, Spergon Technical, Tetrachloro-1,4-benzoquinone, Tetrachloro-p-benzoquinone, Tetrachloro-p-quinone, Tetrachlorobenzoquinone, Tetrachloroquinone, Vulklor, p-Chloranil, α-Chloranil
InChI Formula InChI=1S/C6Cl4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11
SMILES O=C1C(Cl)=C(Cl)C(=O)C(Cl)=C1Cl
IUPAC Name 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 245.88
Formula Weight 243.8652
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.