missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloro-1,4-benzoquinone, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Chloranil |
| CAS | 118-75-2 |
| Molecular Formula | C6 Cl4 O2 |
| Quantity | 5 g |
| Synonym | p-Benzoquinone','2,3,5,6-tetrachloro- (8CI), 2,3,5,6-Tetrachloro-2,5-cyclohexadiene-1,4-dione, 1,2,4,5-Tetrachlorobenzoquinone, 2,3,5,6-Tetrachloro-1,4-(p-)-benzoquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 2,3,5,6-Tetrachloro-p-benzoquinone, 2,3,5,6-Tetrachlorobenzoquinone, 2,3,5,6-Tetrachlorocyclohexadiene-1,4-dione, 2,3,5,6-Tetrachloroquinone, Actor CL, Chloranil, Coversan, Dow Seed Disinfectant No. 5, ENT 3797, NSC 8432, Psorisan, Quinone tetrachloride, Reranil, Spergon, Spergon I, Spergon Technical, Tetrachloro-1,4-benzoquinone, Tetrachloro-p-benzoquinone, Tetrachloro-p-quinone, Tetrachlorobenzoquinone, Tetrachloroquinone, Vulklor, p-Chloranil, α-Chloranil |
| InChI Formula | InChI=1S/C6Cl4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11 |
| SMILES | O=C1C(Cl)=C(Cl)C(=O)C(Cl)=C1Cl |
| IUPAC Name | 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 245.88 |
| Formula Weight | 243.8652 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction