CAS RN 1049-84-9
CAS RN 1049-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4S)-2-[Carboxy[(phenoxy-acetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Phenoxymethylpenicillin), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.