CAS RN 1040085-98-0
CAS RN 1040085-98-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.