Specifications
| Chemical Name or Material | Zopiclone |
| CAS | 43200-80-2 |
| Quantity | 1 g |
| Solubility | soluble in acetone, chloroform, DMSO |
| Merck Index | 10245 |
| Melting Point | 173°C to 175°C |
| Molecular Formula | C17 H17 Cl N6 O3 |
| Formula Weight | 388.81 |
| Physical Form | Powder |
| Color | White to Light-yellow |