Learn More
Zolmitriptan, TRC
Product Code. 30003331
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003331 1 g 1g
30137428 50 mg 50mg
30003928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003331 Supplier Toronto Research Chemicals Supplier No. TRCZ6390001G
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 139264-17-8
Molecular Formula C16 H21 N3 O2
Molecular Weight (g/mol) 287.36
Synonym Zolmitriptan, (4S)-4-[[3-[2-Dimethylamino)ethyl]-1H-indol-5-yl]methyl]oxazolidin-2-one, (S)-4-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-2-oxazolidinone, 311C90, Asco Top, BW 311C90, Xolnox, Zipton, Zomig, Zominat
IUPAC Name (4S)-4-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one
SMILES CN(C)CCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H21N3O2/c1-19(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-21-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1
Formula Weight 287.1634
Chemical Name or Material Zolmitriptan