Learn More
Novus Biologicals™ ZNF554 293T Cell Transient Overexpression Lysate
Product Code. 18670311 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18670311 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18670311 Supplier Novus Biologicals™ Supplier No. H00115196T01
 Restricted Item
Estimated Shipment: 17-08-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target ZNF554
Synonym FLJ34817, zinc finger protein 554
Gene Symbol ZNF554