Learn More
RETSCH Zirconium Oxide Grinding Balls
Product Code. 11984382 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
Cell and Tissue Disruption with 200,400 mm
Centrifugal Ball Mill S 100
High Energy Ball Mill Emax
MM 200 and MM 301
MM 200 and MM 400
MM 400
Diameter (Metric):
0.1 mm
0.5 mm
1 mm
2 mm
3 mm
5 mm
12 mm
15 mm
40 mm
Unit Size:
Each
Pack of 200
Product Code. 11984382
Brand: RETSCH 32.368.0004
Only null left
Specifications
Product Type Grinding Balls
Diameter (Metric) 1 mm
For Use With (Equipment) High Energy Ball Mill Emax
Material Zirconium Oxide