Learn More
RETSCH Zirconium Oxide Grinding Balls
Product Code. 10216702 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
Cell and Tissue Disruption with 200,400 mm
Centrifugal Ball Mill S 100
High Energy Ball Mill Emax
MM 200 and MM 301
MM 200 and MM 400
MM 400
Diameter (Metric):
0.1 mm
0.5 mm
1 mm
2 mm
3 mm
5 mm
12 mm
15 mm
40 mm
Unit Size:
Each
Pack of 200
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Unit Size
10216702 MM 400 5 mm Pack of 200
10354671 MM 200 and MM 301 12 mm Each
10608751 Cell and Tissue Disruption with 200,400 mm 2 mm Each
10226322 MM 200 and MM 400 15 mm Each
10492262 MM 400 3 mm Pack of 200
10419112 Centrifugal Ball Mill S 100 40 mm Each
11964382 High Energy Ball Mill Emax 0.5 mm Each
11984382 High Energy Ball Mill Emax 1 mm Each
11974382 High Energy Ball Mill Emax 0.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10216702 Supplier RETSCH Supplier No. 22.455.0009
  • ONLINE EXCLUSIVE PRICE
    275.00€
    254.81€ / Pack of 200
    Save 20.19€
    7% Off
Estimated Shipment: 20-01-2027