Learn More
RETSCH Zirconium Oxide Aeration Lids
1901.28€ - 5160.00€
Specifications
Product Type | Aeration Lid |
---|---|
Closure Type | Aeration Lid |
Material | Zirconium Oxide |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
15330964
|
RETSCH
01.107.0569 |
Zirconium Oxide |
5160.00€
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
11914382
|
RETSCH
22.107.0010 |
Zirconium Oxide |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15300964
|
RETSCH
01.107.0564 |
Zirconium Oxide |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Aeration Lid | |
Aeration Lid | |
Zirconium Oxide |