Learn More
RETSCH Zirconium Oxide Aeration Lids
Product Code. 15330964 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
125 mL Emax Grinding Jar
50 mL Emax Grinding Jar
500 mL Grinding Jar
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15330964 50 mL Emax Grinding Jar Each
11914382 500 mL Grinding Jar Each
15300964 125 mL Emax Grinding Jar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15330964 Supplier RETSCH Supplier No. 01.107.0569
  • ONLINE EXCLUSIVE PRICE
    5520.00€
    4662.84€ / Pack of 1
    Save 857.16€
    15% Off
Estimated Shipment: 27-01-2027
Specifications
Product Type Aeration Lid
Closure Type Aeration Lid
For Use With (Equipment) 50 mL Emax Grinding Jar
Material Zirconium Oxide