Learn More
Thermo Scientific™ Zirconium Crucible
Product Code. 15433605
Click to view available options
Diameter (Metric) Outer:
39 mm
42 mm
47 mm
Height (Metric):
33 mm
35 mm
36 mm
44 mm
Description:
Flanged Rim OD: 44 mm
Flanged Rim OD: 48 mm
Flanged Rim OD: 54 mm
SlideOn Ring OD: 48 mm
SlideOn Ring OD: 54 mm
SnapOn Ring OD: 44 mm
SnapOn Ring OD: 48 mm
Snapon Ring OD: 50mm
Unit Size:
Each
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15433605
Brand: Thermo Scientific™ 035966.KT
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Description Snapon Ring OD: 50mm
Material Zirconium
Diameter (Metric) Outer 47 mm
Height (Metric) 36 mm
Product Type Zirconium Crucible
Diameter (Metric) Base 38mm