Learn More
Chemsplash Xtreme 50 SMS Coverall Type 5/6 Disposable

Description
Chemsplash Xtreme SMS 50 Coveralls
- 50GSM Breathable SMS Fabric
- Elasticated 3 Pce Hood
- Elasticated Cuffs
- Elasticated Ankles
- Two Way Zip
- Adhesive Zip Flap
- Silicone and Latex Free
- See style 2544 for White Sizes S-3XL
- Comes without the chest label
Description: Chemsplash Xtreme SMS 50 disposable coveralls are made from extremely breathable SMS fabric, suitable for prolonged periods of use. Featuring an elasticated hood, cuffs, and ankles, with a two-way zip and an adhesive cover flap. Its strong and robust properties provide a good chemical barrier, especially to dry hazardous particles, while being extremely comfortable to wear, offering the best value within our Cat III Type 5 and 6 range.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | 4X-Large |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | Tiger Stripe |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.