missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Dihydrogen Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Zinc Dihydrogen Phosphate |
| CAS | 13598-37-3 |
| Molecular Formula | ZnH4P2O8 |
| Quantity | 2.5 g |
| Synonym | Zinc phosphate (Zn(H2PO4)2) (6CI','7CI), Budit T 21, Monobasic zinc phosphate, Z 21-82, Zinc bis(dihydrogen phosphate), Zinc diacid phosphate, Zinc dihydrogen phosphate, Zinc dihydrogen phosphate (Zn(H2PO4)2), Zinc dihydrophosphate, Zinc hydrogen phosphate (Zn(H2PO4)2), Zinc phosphate (1:2), Zinc primary phosphate |
| InChI Formula | InChI=1S/2H3O4P.Zn/c2*1-5(2,3)4;/h2*(H3,1,2,3,4);/q;;+2/p-2 |
| SMILES | OP(O)([O-])=O.OP(O)([O-])=O.[Zn+2] |
| IUPAC Name | zinc(II) dihydrogen phosphate |
| Molecular Weight (g/mol) | 259.35 |
| Formula Weight | 257.87 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?