Learn More
Usbeck Laborgeraete™ Zinc Die-casting Stand Clamps
Click to view available options
Size (Metric) Grip:
12 to 25 mm
20 to 40 mm
30 to 80 mm
38 to 60 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | Each |
| Type | Stand Clamp |
| Material | Zinc Die Cast, Powder Coated |
| Size (Metric) Grip | 30 to 80 mm |