missing translation for 'onlineSavingsMsg'
Learn More

Zinc cyanide, 98%

Product Code. 11456840
Click to view available options
Quantity:
25 g
100 g
1000 g
Unit Size:
1000g
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 11456840

Brand: Thermo Scientific Alfa Aesar L15836.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 557-21-1 | Zn(CN)2

Zinc cyanide is used for electroplating of zinc. In Gatterman reaction , it is used for the introduction of formyl group in to aromatic compounds as well as to prepare 2-hydroxy-1-nafthaldehyde and mesitaldehyde. It is also used in chemical analysis in analytical chemistry. Further, it acts as a catalyst for the cyanosilylation of aldehydes and ketones. In addition to this, it reacts with 1,3,5-triethyl-benzene to get 2,4,6-triethyl-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc cyanide is used for electroplating of zinc. In Gatterman reaction , it is used for the introduction of formyl group in to aromatic compounds as well as to prepare 2-hydroxy-1-nafthaldehyde and mesitaldehyde. It is also used in chemical analysis in analytical chemistry. Further, it acts as a catalyst for the cyanosilylation of aldehydes and ketones. In addition to this, it reacts with 1,3,5-triethyl-benzene to get 2,4,6-triethyl-benzaldehyde.

Solubility
Soluble in alkalies, potassium cyanide and ammonia. Insoluble in water and most solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-21-1
Molecular Formula Zn(CN)2
MDL Number MFCD00011292

Specifications

Melting Point 800°C (decomposition)
Quantity 1000 g
Linear Formula Zn(CN)2
UN Number UN1713
Beilstein 4124366
Merck Index 14,10135
Solubility Information Soluble in alkalies,potassium cyanide and ammonia. Insoluble in water and most solvents.
Formula Weight 117.42
Percent Purity 98%
Sensitivity Hygroscopic
Density 1.852
Chemical Name or Material Zinc cyanide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.