Learn More
ZVG™ zetPutz™ Fleece Cloths
Description
Highly absorbent specialty cotton fabric based for versatile cleaning and maintenance tasks. In the zetPutz™ range, you will find additional universal cloths, sponges, and dishcloths for other cleaning tasks. When small as well as coarse soiling needs to be removed effectively and economically, ZVG™ products are ideal.
- Universally applicable
- Highly absorbent
- Low-lint
- Tear-resistant
Specifications
Specifications
| Product Type | Cloth |
| Length (English) | 13.8 in. |
| Length (Metric) | 35 cm |
| Width (English) | 13 in. |
| Width (Metric) | 33 cm |
| Material | Fleece |
| Color | White |
| Disposable | Reusable |
| Packaging Quantity | 415 Wipes/Pk. |
| Sterility | Non-sterile |