Learn More
ZVG™ zetMedica™ Medical Tissue Rolls
Product Code. 10737534
Change view
Click to view available options
Weight (Metric):
10.26 kg
10.59 kg
Width (Metric):
55 cm
59 cm
Unit Size:
9 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Width (Metric) Unit Size
10737534 10.59 kg 55 cm 9 rolls
10549672 10.26 kg 59 cm 9 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10737534 Supplier ZVG™ Supplier No. 1225500
  • ONLINE EXCLUSIVE PRICE
    48.24€
    35.36€ / 9 rolls
    Save 12.88€
    26% Off
Estimated Shipment: 19-01-2027
This perforated medical roll tissue features dot-to-dot embossing for easy handling. The white tissue material ensures effective and hygienic use.
  • Effective in quickly preparing the premises in the care area for the next patient
  • Tissue is soft and highly absorbent
  • Material does not stick to the skin, and immediately absorbs moisture and grease
  • Different rolls are economically usable thanks to their perforation and always remain clean at the tear-off edge
  • Suitable doctor's roll holder can be mounted either on the wall or directly on the bed
  • Roll contains approximately 130 tear-off sheets
  • Made with point-to-point embossing
In a novel manufacturing process, the individual layers of the cellulose cloth are bonded together at many points across the entire surface. This creates a strong bond between the layers, making them highly tear-resistant. This process combines the advantages of a thick wiping cloth (stability) with those of a multi-layered cloth (high absorbency).
Specifications
Product Type Roll Paper
Color White
Material Paper
Sterility Non-sterile
Disposable Single-use
Length (Metric) 50 m
Ply 2-ply
Quantity 9 Rolls/Pk.
Weight (Metric) 10.59 kg
Width (Metric) 55 cm