Learn More
ZEOprep™ SH (Thiol) Functionalized
Shop All Zeochem Products
Click to view available options
:
1 kg
100 g
5 kg
500 g
Description
- SH (Thiol) functionalized for Ion Exchange separations
- Idea for weak anion applications
- Irregular Amorphous Silica Gel
- High Purity
- European Manufacture
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- High Chemical and Mechanical Stability
- Narrow Particles Size Distribution
- 20 to 500 μm particles size range
- Available in 60 A or 90 A pore size
- High loadability
- Selection of pack sizes available
- Suitable for gravity, flash, prep and process scale
Specifications
Specifications
| Particle Size | 40 to 63 μm |
| Pore Size | 60 Å |
| Technique | Chromatography |
| Quantity | 1 kg |
| Type | Silica Gel |
| Material | Silica Gel |
| Phase | SH (Thiol) Functionalized |
| Particle Shape | Irregular |